Learn More
Medchemexpress LLC Justicidin A 1mg | 25001-57-4 | 394.37 | C22H18O7 | 1 MG

Supplier: Medchemexpress LLC HY1170781MG
Justicidin A is a naturally occurring lignan used as a research reagent with reported apoptosis-inducing activity in cellular studies. Supplied as a solid for dissolution in organic solvents, it is intended for in vitro biochemical and cell biology research applications.
- High purity: 99.63%.
- Molecular formula C22H18O7 and molecular weight 394.37.
- Soluble in DMSO at 16.67 mg/mL (heat and sonicate as needed).
- Storage: store at 4°C; in solution store at -80°C for up to 6 months or -20°C for up to 1 month; protect from light.
- Available in small research quantities (1 mg, 5 mg, 10 mg).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.