Learn More
Medchemexpress LLC JW 642 | 1416133-89-5 | MFCD22987956 | 99.9% | 462.39 g/mol | C21H20F6N2O3 | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY1233210MM 1ML
Encompass_Preferred
JW 642 is a potent, selective monoacylglycerol lipase (MAGL) inhibitor used in biochemical and pharmacological research. It is supplied as a solution in DMSO and as solid quantities for assay and in vivo studies. Reported IC50 values include 3.7 nM (human), 7.6 nM (mouse), and 14 nM (rat), demonstrating high potency across species.
Catalog No. 50-002-39061