Learn More
Medchemexpress LLC JWG-071 | 2250323-50-1 | 99.2% | 612.76 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY10888610MG
Encompass_Preferred
JWG-071 is identified as a kinase-selective chemical probe for ERK5. It demonstrates inhibitory activity against both ERK5 and LRRK2, with IC50 values of 88 nM and 109 nM, respectively. The product is intended for research use only and is not sold to patients.
  • Kinase-selective chemical probe for ERK5.
  • Inhibits ERK5 with an IC50 of 88 nM.
  • Inhibits LRRK2 with an IC50 of 109 nM.
  • Available in solid and solution forms for various experimental needs.
  • Detailed solubility and storage information provided for optimal product handling.
  • Associated with publications in scientific journals, indicating its use in research.
Catalog No. 50-003-46332