Learn More
Cayman Chemical JWH 018 2naphthylN 2 2dI 5mg
SDP
Supplier:  Cayman Chemical 90010085 MG
Encompass_Preferred
Differs structurally from JWH 018 by having the naphthyl group attached at the 2' position and 2,2-dimethylpropyl in place of the pentyl chain
Catalog No. 50-289-4978