Learn More
Cayman Chemical JWH 018 N 2 2dImethylprpyl 5mg
SDP
Supplier:  Cayman Chemical 90010015 MG
Encompass_Preferred
Differs from JWH 018 structurally by having a 2,2-dimethylpropyl group, rather than a pentyl chain, extending from the indole group; intended for research and forensic applications
Catalog No. 50-289-3005