Learn More
Cayman Chemical JWH 018 N- 5-hydroxypentyl10mg
SDP
Supplier:  Cayman Chemical 1095810 MG
Encompass_Preferred
An analytical reference standard categorized as a synthetic cannabinoid; a major urinary metabolite of JWH 018; intended for research and forensic applications
Catalog No. 50-295-1426