Learn More
Cayman Chemical JWH 018 N 5hydroxypentyl m
SDP
Supplier:  Cayman Chemical 10933500 UG
Encompass_Preferred
An internal standard for the quantification of JWH 018 N-(5-hydroxypentyl) metabolite by GC- or LC-MS
Catalog No. 50-295-1173