Learn More
Cayman Chemical JWH 073 2naphthylN 1meth 10mg
SDP
Supplier:  Cayman Chemical 900101610 MG
Encompass_Preferred
Differs structurally from JWH 073 by having the naphthyl group attached at the 2' position and a 1-methylpropyl group in place of a butyl chain
Catalog No. 50-289-5727