Learn More
Cayman Chemical JWH 073 N 3hydroxybutyl 100ug
SDP
Supplier:  Cayman Chemical 10927100 UG
Encompass_Preferred
An internal standard for the quanitification of (±)-JWH 073 N-(3-hydroxybutyl) metabolite by GC- or LC-MS
Catalog No. 50-295-1223