Learn More
Cayman Chemical JWH 081 NpentnoIc ACD meta 1mg
SDP
Supplier:  Cayman Chemical 143621 MG
Encompass_Preferred
An internal standard for the quantification of JWH 081 N-pentanoic acid metabolite by GC- or LC-MS
Catalog No. 50-295-3916