Learn More
Cayman Chemical JWH 145 Analytical Reference S
SDP
Supplier:  Cayman Chemical 108251 MG
Encompass_Preferred
A (1-naphthoyl)pyrrole analog of JWH 018 that potently activates both CB1 and CB2 receptors (Ki values of 14 and 6.4 nM, respectively); intended for forensic and research applications
Catalog No. 50-288-7924