Learn More
Cayman Chemical JWH 203 3CLOrophenl IsomR 10mg
SDP
Supplier:  Cayman Chemical 1086810 MG
Encompass_Preferred
An analytical reference standard that is structurally similar to known synthetic cannabinoids; intended for research and forensic applications
Catalog No. 50-295-1631