Learn More
Cayman Chemical JWH 210 3ethylnphthyl ISME 5mg
SDP
Supplier:  Cayman Chemical 90010395 MG
Encompass_Preferred
A positional isomer of JWH 210, having the ethyl side chain at the 3 position rather than at the 4 position of the naphthyl group; intended for forensic purposes
Catalog No. 50-289-5160