Learn More
Cayman Chemical JWH 250 5hydroxyIndl meta 1mg
SDP
Supplier:  Cayman Chemical 90007661 MG
Encompass_Preferred
Expected to be a metabolite of JWH 250 that would be detectable both in serum and in urine; intended for research and forensic applications
Catalog No. 50-289-5981