Learn More
Cayman Chemical JWH 251 3methylphenyl IsmR 5mg
SDP
Supplier:  Cayman Chemical 90010215 MG
Encompass_Preferred
Differs from JWH 251 by having the methyl group at the 3 position, rather than the 2 position, on the phenyl group
Catalog No. 50-289-2970