Learn More
Medchemexpress LLC JX06 | 729-46-4 | 99.78% | 324.51 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1956410MM/1ML
Encompass_Preferred
JX06 is a potent, selective, and covalent inhibitor of Pyruvate Dehydrogenase Kinase (PDK), inhibiting PDK1, PDK2, and PDK3 with varying IC50s. It functions by irreversibly binding to a cysteine residue in PDK1 and has demonstrated significant antitumor activity in both in vitro and in vivo studies.
  • Potent, selective, and covalent PDK inhibitor.
  • Inhibits PDK1, PDK2, and PDK3.
  • Induces cell apoptosis in various cancer cell lines.
  • Suppresses cancer cell growth.
  • Inhibits PDHA1 phosphorylation.
  • Increases glucose uptake and intracellular ATP.
  • Reduces aerobic glycolysis.
  • Induces ROS generation in cancer cells.
  • Demonstrates in vivo tumor growth inhibition.
Catalog No. 50-004-00555