Learn More
Medchemexpress LLC Jyq-164 | 99.4% | 530.62 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1635625MG
Encompass_Preferred
JYQ-164 is a small molecule inhibitor of Parkinson's disease protein 7 (PARK7/DJ-1) in humans. JYQ-164 works by covalently and selectively targeting Cys106, a key residue of PARK7 (IC50=21 nM). The high inhibitory effect of JYQ-164 on PARK7 is 5 times more potent than the previously reported inhibitor JYQ-88. JYQ-164 can be used in Parkinson's disease and cancer research.
Catalog No. 50-003-94803