Learn More
Abcam JZL-184, monoacylglycerol lipase (MAGL) inhibitor, 10MG
Supplier:  Abcam AB14159210MG
Encompass_Preferred
MW 520.5 Da, Purity >98%. Potent, selective and irreversible inhibitor of MAGL (IC₅₀ values are 8 nM and 4 μM for MAGL and FAAH respectively). Cell-permeable. Potentiates DSE in cerebellar Purkinje neurons and DSI in hippocampal CA1 pyramidal neurons. Inhibits nociceptive processing and central sensitization in vivo..
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6503