Learn More
Apexbio Technology LLC JZL184 1101854-58-3 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B19585.1
Encompass_Preferred
JZL184 (CAS 1101854-58-3) is a potent and selective inhibitor of monoacylglycerol lipase (MAGL) a serine hydrolase that hydrolyzes the endocannabinoid 2-arachidonoylglycerol (2-AG) In murine models JZL184 inhibits MAGL-mediated 2-AG hydrolysis with an IC50 of 8 nM resulting in elevated brain 2-AG concentrations Electrophysiological studies demonstrate that JZL184 modulates synaptic plasticity in cerebellar and hippocampal neurons via CB1 receptor-dependent mechanisms such as delayed depolarization-induced suppression of inhibition and excitation In vivo JZL184 elicits CB1-dependent behaviors including analgesia and hypothermia and exhibits anxiolytic and antinociceptive effects in rodent models This compound is widely used to investigate endocannabinoid signaling and MAGL function in neurobiological research
Catalog No. 50-000-92918