Learn More
TARGETMOL CHEMICALS INC K-111 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T277061MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. K-111 (BM-170744) is an orally available peroxisome proliferator-activated receptor (PPAR)-alpha agonist and insulin enhancer for the study of obesity-type diabetes mellitus and the metabolic syndrome. Purity 99.93%
Catalog No. 50-287-9424