Learn More
Medchemexpress LLC K-252a | 99533-80-9 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN673210MM/1ML
Encompass_Preferred
K-252a is a staurosporine analog that functions as a potent protein kinase inhibitor. It selectively inhibits various kinases including PKC, PKA, Ca2+/calmodulin-dependent kinase type II, and phosphorylase kinase. This compound also acts as a powerful inhibitor of the tyrosine protein kinase (TRK) activity associated with the NGF receptor gp140trk, effectively inhibiting NGF-promoted neurite outgrowth in in vitro studies.
  • Inhibits a wide range of protein kinases.
  • Acts as a potent inhibitor of TRK activity.
  • Effectively inhibits NGF-promoted neurite outgrowth.
  • Useful for research involving kinase activity and neurite development.
Catalog No. 50-004-19469