Learn More
Medchemexpress LLC K-252a | 99533-80-9 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN673210MG
Encompass_Preferred
K-252a is a staurosporine analog that acts as a protein kinase inhibitor. It exhibits specific inhibitory activity against PKC, PKA, Ca2+/calmodulin-dependent kinase type II, and phosphorylase kinase. Additionally, it is a potent inhibitor of the tyrosine protein kinase (TRK) activity of the NGF receptor gp140trk.
  • Inhibits protein kinase
  • Targets PKC, PKA, CaMK, autophagy, Trk receptor, and acts as an antibiotic
  • Involved in pathways such as epigenetics, TGF-beta/Smad, stem cell/Wnt, neuronal signaling, autophagy, protein tyrosine kinase/RTK, and anti-infection
Catalog No. 50-004-18311