Learn More
Medchemexpress LLC K-252a | 99533-80-9 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN67325MG
Encompass_Preferred
K-252a is a staurosporine analog that inhibits protein kinase with IC50 values of 470 nM for PKC, 140 nM for PKA, 270 nM for Ca2+/calmodulin-dependent kinase type II, and 1.7 nM for phosphorylase kinase. It is also a potent inhibitor (IC50 of 3 nM) of the tyrosine protein kinase (TRK) activity of the NGF receptor gp140trk, which is a product of the trk protooncogene.
  • A staurosporine analog.
  • Inhibits protein kinase C (PKC) with an IC50 of 470 nM.
  • Inhibits protein kinase A (PKA) with an IC50 of 140 nM.
  • Inhibits Ca2+/calmodulin-dependent kinase type II with an IC50 of 270 nM.
  • Inhibits phosphorylase kinase with an IC50 of 1.7 nM.
  • Potent inhibitor of the tyrosine protein kinase (TRK) activity of the NGF receptor gp140trk with an IC50 of 3 nM.
  • Appears as a white to light yellow solid.
Catalog No. 50-004-19328