Learn More
TARGETMOL CHEMICALS INC K-7174 dihydrochloride 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T11742L10MG
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg and bulk. Please contact Fisher for quotes. K-7174 dihydrochloride is a proteasome and GATA inhibitor that inhibits neuronal degeneration and induces apoptosis. Purity 99.15%
Catalog No. 50-288-0696