Learn More
Medchemexpress LLC K-756 | 130017-40-2 | 433.50 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYU00422100MG
Encompass_Preferred
K-756 is a direct and selective tankyrase (TNKS) inhibitor that inhibits the ADP-ribosylation activity of TNKS1 and TNKS2. This novel and selective Wnt/β-catenin pathway inhibitor binds to the induced pocket of TNKS, inhibiting its enzyme activity and showing isoform selectivity.
  • Direct and selective tankyrase (TNKS) inhibitor
  • Inhibits ADP-ribosylation activity of TNKS1 and TNKS2
  • Novel and selective Wnt/β-catenin pathway inhibitor
  • Binds to the induced pocket of TNKS
  • Inhibits cell growth in APC-mutant colorectal cancer cells
Catalog No. 50-004-48942