Learn More
Medchemexpress LLC K-8012 | 1346513-17-4 | 98.2% | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1214355MG
Encompass_Preferred
K-8012 is a small-molecule RXRα antagonist used for research. It has molecular formula C23H23FN4, molecular weight 374.45 g/mol, and reported purity of 98.24%.
  • Antagonist of retinoid X receptor alpha (RXRα).
  • Reported activity in PI3K/AKT and apoptosis pathways.
  • High purity suitable for research applications.
  • Available as a 5 mg research-grade sample.
Catalog No. 50-002-31032