Learn More
TARGETMOL CHEMICALS INC K-975 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T99545MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. K-975 is a highly selective, orally active TEAD inhibitor, effectively inhibiting protein-protein interactions between TEAD and YAP1/TAZ.
Catalog No. 50-287-5899