Learn More
Cell Biolabs Inc K-Ras Activation Assay Kit, 20 assays
Supplier:  Cell Biolabs Inc STA400K
Encompass_Preferred
Cell Biolabs’ K-Ras Activation Assay Kit provides a simple and fast tool to monitor the activation of K-Ras. The kit utilizes easily identifiable Raf1 RBD Agarose beads (pink in color) to selectively isolate and pull-down the active form of Ras from purified samples or endogenous lysates. Subsequently, the precipitated GTP-Ras is detected by western blot analysis using an Anti-K-Ras specific polyclonal antibody. Each kit includes an Immunoblot Positive Control and provides sufficient quantities to perform 20 assays.
Catalog No. NC0472675
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.