Learn More
Santa Cruz Biotechnology K-RAS (F234) MOUSE MONOCL
Supplier:  Santa Cruz Biotechnology SC30
Encompass
K-Ras (F234), Mouse monoclonal IgG 2a, 200ug/mL, Raised against recombinant Ras Protein encompassing amino acid residues 54 to 189, Recommended for detection of K-Ras p21 of M, R and H origin by WB, IP and IF
Catalog No. NC9903161