Learn More
Medchemexpress LLC K-Ras(G12C) inhibitor 12 | 1469337-95-8 | 99.6% | 449.67 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1870710MM/1ML
Encompass_Preferred
K-Ras(G12C) inhibitor 12 is an irreversible inhibitor of K-Ras(G12C). It can alter the nucleotide-binding preference of K-Ras and block its interaction with effector proteins. This inhibitor can reduce cell viability and induce apoptosis in lung cancer cell lines with G12C mutations, demonstrating anti-tumor activity.
  • Irreversible inhibitor
  • Alters nucleotide-binding preference of K-Ras
  • Blocks interaction with effector proteins
  • Reduces cell viability in lung cancer cell lines with G12C mutations
  • Induces apoptosis in lung cancer cell lines with G12C mutations
  • Has anti-tumor activity
Catalog No. 50-003-85707