Learn More
Cayman Chemical K-Ras G12C InhIbItr 12 5mg
SDP
Supplier:  Cayman Chemical 201185 MG
Encompass_Preferred
An allosteric inhibitor of oncogenic K-Ras(G12C); decreases viability and increases apoptosis in several lung cancer cell lines that contain G12C mutations (EC50 = 0.32 µM in H1792 cells)
Catalog No. 50-298-3772