Learn More
AdipoGen K-TCPB
SDP
Supplier:  AdipoGen CDXP0134G001
Encompass_Preferred
Chemical. CAS 14680-77-4. Formula C24H16BCl4K. MW 496.1. Synthetic. Lipophilic anionic additive, used together with selective ionophores. The ionophore is the ion carrier which reversibly binds ions and transports them across the hydrophobic membrane. The ionic additive K-TCPB, catalyzes the ion-exchange process at the sample/membrane interface.
Catalog No. 50-256-0227