Learn More
Med Vet International Steel Stainless K-Wire, J0250MA, 0.035in x 7in, Each
Supplier:  Med Vet International JOR250MA
Encompass_Preferred
Highest quality stainless steel type 316LVM. U.S.A. made. Double Trocar Point.
Catalog No. 50-005-83106