Learn More
Med Vet International Steel Stainless K-Wire, Single, J0250NA, 0.045in x 7in, Each
Supplier:  Med Vet International JOR250NA
Encompass_Preferred
K-Wire or Fine Orthopedic Wire. Highest quality stainless steel type 316LVM. U.S.A. made. Double Trocar Point.
Catalog No. 50-005-82541