Learn More
Medchemexpress LLC K11 | 1398109-79-9 | 99.90% | 2493.09 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP57041MG
Encompass_Preferred
K11 is an antimicrobial peptide that is active against MDR/XDR K. pneumoniae isolates and inhibits bacterial biofilm formation. It exhibits high thermal and wide pH stability. K11 can act synergistically with various antibiotics such as Chloramphenicol, Meropenem, and Rifampicin against drug-resistant K. pneumoniae.
  • Active against MDR/XDR K. pneumoniae isolates
  • Inhibits bacterial biofilm formation
  • Exhibits high thermal and wide pH stability
  • Can act synergistically with various antibiotics
Catalog No. 50-003-99077