Learn More
Grainger K4796 Silicone Round Cord 5/8 D 25 L 70A Blue
Supplier:  Grainger 16Z178
Encompass_Preferred
The Silicone Round Cord Metal is a silicone cord with a metal core that is designed for high temperature applications up to 500°F. It has a round cross-section and is available in multiple diameters from 1/8 inch to 1 inch. The metal core provides strength while the silicone jacket offers flexibility. This high quality silicone cord is UL recognized and RoHS compliant. With its high temperature resistance and durability, it's well-suited for applications like ovens, heaters, and other thermal equipment. The cord comes in standard 100 foot spools.
Catalog No. 50-276-7536