Learn More
ATCC K-562-GFP; Leukemia(CML)-GFP;Human;(Homo sapiens)
Supplier:  ATCC CCL243GFP
Encompass
K-562-GFP cells are lymphoblasts isolated from the bone marrow of a 53-year-old chronic myelogenous leukemia patient. The K-562-GFP can be used in immune system disorder and immunology research.
Catalog No. 50-238-4422