Learn More
Medchemexpress LLC Kaempferitrin | 482-38-2 | 578.52 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0628
Encompass_Preferred
Kaempferitrin is a natural flavonoid known for its diverse biological activities. It possesses antinociceptive, anti-inflammatory, anti-diabetic, antitumoral, and chemopreventive effects, and plays a role in activating the insulin signaling pathway. This compound has demonstrated utility in both in vitro and in vivo studies for various research applications.
  • Activates the insulin signaling pathway.
  • Exhibits antinociceptive, anti-inflammatory, anti-diabetic, antitumoral, and chemopreventive effects.
  • Increases insulin receptor beta tyrosine phosphorylation and stimulates Akt phosphorylation.
  • Promotes GLUT4 translocation and increases total Glu4 protein levels.
  • Cytotoxic to human cancer cells like HeLa and MDA-MB231.
  • Induces apoptosis in cancer cells.
  • Suppresses tumor growth and inhibits cell proliferation in vivo.
Catalog No. 50-003-89892