Learn More
Medchemexpress LLC Kaempferol 3-neohesperidoside | 32602-81-6 | MFCD20261019 | 99.7% | 594.52 g·mol⁻¹ | C27H30O15 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10720725MG
Encompass_Preferred
Kaempferol 3-O-neohesperidoside is a flavonoid glycoside (C27H30O15, MW 594.52 g·mol⁻¹) supplied as a light yellow solid. It has reported insulinomimetic activity in rat soleus muscle and is intended for laboratory research use only.
  • High purity (99.7%) for analytical and biological studies.
  • Flavonoid glycoside with reported insulinomimetic activity.
  • Available as solid and as 10 mM solution in DMSO for ready use.
  • Chemical identity: formula C27H30O15, molecular weight 594.52 g·mol⁻¹.
  • Light yellow solid appearance suitable for storage and handling in research labs.
Catalog No. 50-002-41070