Learn More
Medchemexpress LLC HY-N7176 1mg Medchemexpress, Kaempferol 3-O-β-D-glucuronide CAS:22688-78-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN71761MG
Encompass_Preferred
Medchemexpress, HY-N7176 1mg Kaempferol 3-O-β-D-glucuronide CAS:22688-78-4 Kaempferol 3-O-β-D-glucuronide (Kaempferol-3-glucuronide), one conjugated kaempferol metabolite, has anti-inflammatory effect. Kaempferol 3-O-β-D-glucuronide significantly inhibits various pro-inflammatory mediators like IL-1β, NO, PGE2, and LTB4. Kaempferol 3-O-β-D-glucuronide upregulates the secretion of anti-inflammatory cytokine IL-10. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2990