Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Kaempferol 3-O-rutinoside 7-O-glucoside | 34336-18-0 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYN81655MG

Encompass_Preferred

Kaempferol 3-O-rutinoside 7-O-glucoside is a flavonoid glycoside found in red tomato, as well as organisms such as Crocus minimus and Crocus chrysanthus. It is intended for research use only.

  • Purity: 99.67%
  • Molecular formula: C33H40O20
  • Molecular weight: 756.66
  • Appearance: Off-white to light yellow solid
  • Classified as a flavonoid, flavonol, phenol, and polyphenol

Catalog No. 50-004-17606


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.