Learn More
Medchemexpress LLC Kaempferol-7,4'-dimethyl ether | 15486-33-6 | 314.29 | 5 MG
Supplier: Medchemexpress LLC HYN17895MG
Kaempferol-7,4'-dimethyl ether is a chemical compound known for its biological activity as a PTP1B inhibitor. It also demonstrates the ability to inhibit the production of nitric oxide (NO).
- PTP1B inhibitor
- Inhibits nitric oxide (NO) production
- Shows an inhibition rate of 46.1% at 100 μM
- Inhibits PTP1B activity with an IC50 value of 16.92 μM