Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kahle's Solution, Ricca Chemical
SDP

$353.00 - $975.00

Specifications

Chemical Name or Material Kahle's Solution
Color Colorless Liquid
CAS 7732-18-5 , 64-17-5 , 64-19-7 , 50-00-0 , 67-56-1 , 67-63-0
CAS Max % 47.29, 32.07, 11.27, 4.26, 3.33, 1.77
Grade Laboratory
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
R42400001A
SDP
View Documents
Ricca Chemical Company
R42400001A
1 L
Each for $353.00
Add to Cart
 
R42400004A
SDP
View Documents
Ricca Chemical Company
R42400004A
4 L
Each for $975.00
Add to Cart
 
Description

Description

Specifications

Specifications

Kahle's Solution
7732-18-5 , 64-17-5 , 64-19-7 , 50-00-0 , 67-56-1 , 67-63-0
Laboratory
Natural Poly Bottle
Actual Value Reported
Colorless Liquid
47.29, 32.07, 11.27, 4.26, 3.33, 1.77
Pass
Actual Value Reported
Non-sterile
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.