CAS RN 67-56-1
- (14)
- (129)
- (2)
- (647)
- (1)
- (3)
- (12)
- (1)
- (1)
- (1)
- (4)
- (1)
- (1)
- (1)
- (3)
- (4)
- (35)
- (1)
- (27)
- (16)
- (1)
- (2)
- (3)
- (1)
- (4)
- (1)
- (101)
- (3)
- (2)
- (1)
- (12)
- (37)
- (1)
- (1)
- (1)
- (4)
- (1)
- (1)
- (56)
- (1)
- (1)
- (15)
- (1)
- (4)
- (1)
- (1)
- (1)
- (1)
- (3)
- (1)
- (1)
- (1)
- (1)
- (1)
- (1)
- (1)
- (1)
- (1)
- (1)
- (1)
- (2)
- (1)
- (1)
- (1)
- (2)
- (2)
- (1)
- (1)
- (1)
- (1)
- (1)
- (1)
- (1)
- (1)
- (1)
- (1)
- (1)
- (1)
- (1)
- (1)
- (411)
- (1)
- (1)
- (2)
- (1)
- (2)
- (1)
- (1)
- (1)
- (1)
- (2)
- (1)
- (1)
- (3)
- (1)
- (1)
- (2)
- (1)
- (1)
- (2)
- (1)
- (1)
- (1)
- (1)
- (1)
- (1)
- (5)
- (1)
- (1)
- (1)
- (3)
- (1)
- (1)
- (1)
- (1)
- (1)
- (1)
- (1)
- (4)
- (7)
- (1)
- (1)
- (1)
- (1)
- (1)
- (10)
- (69)
- (1)
- (2)
- (8)
- (8)
- (5)
- (1)
- (1)
- (2)
- (4)
- (10)
- (2)
- (5)
- (3)
- (5)
- (3)
- (1)
- (5)
- (3)
- (6)
- (4)
- (3)
- (1)
- (1)
- (33)
- (5)
- (1)
- (7)
- (6)
- (6)
- (24)
- (13)
- (4)
- (2)
- (47)
- (3)
- (1)
- (1)
- (10)
- (3)
- (3)
- (2)
- (1)
- (1)
- (2)
- (6)
- (70)
- (4)
- (4)
- (2)
- (4)
- (2)
- (3)
- (3)
- (1)
- (1)
- (1)
- (7)
- (1)
- (1)
- (1)
- (1)
- (2)
- (1)
- (3)
- (3)
- (1)
- (3)
- (3)
- (34)
- (10)
- (56)
- (7)
- (2)
- (1)
- (9)
- (21)
- (3)
- (1)
- (2)
- (1)
- (1)
- (2)
- (2)
- (1)
- (1)
- (4)
- (1)
- (2)
- (1)
- (2)
- (1)
- (1)
- (4)
- (2)
- (3)
- (1)
- (2)
- (4)
- (2)
- (1)
- (2)
- (1)
- (4)
- (37)
- (21)
- (1)
- (3)
- (1)
- (1)
- (2)
- (1)
- (11)
- (2)
- (12)
- (1)
- (1)
- (1)
- (16)
- (3)
- (16)
- (1)
- (3)
- (3)
- (2)
- (1)
Filtered Search Results
Methanol, 99.9%, for HPLC
CAS: 67-56-1 Molecular Formula: CH4O Molecular Weight (g/mol): 32.04 MDL Number: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Synonym: methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
| PubChem CID | 887 |
|---|---|
| CAS | 67-56-1 |
| Molecular Weight (g/mol) | 32.04 |
| ChEBI | CHEBI:17790 |
| MDL Number | MFCD00004595 |
| SMILES | CO |
| Synonym | methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit |
| IUPAC Name | methanol |
| InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
| Molecular Formula | CH4O |
Methanol ACS, MilliporeSigma™
CAS: 67-56-1 Molecular Formula: CH4O Molecular Weight (g/mol): 32.04 MDL Number: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Synonym: methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
| PubChem CID | 887 |
|---|---|
| CAS | 67-56-1 |
| Molecular Weight (g/mol) | 32.04 |
| ChEBI | CHEBI:17790 |
| MDL Number | MFCD00004595 |
| SMILES | CO |
| Synonym | methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit |
| IUPAC Name | methanol |
| InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
| Molecular Formula | CH4O |
Methanol, OmniSolv Gradient Grade, MilliporeSigma™
CAS: 67-56-1 Molecular Formula: CH4O Molecular Weight (g/mol): 32.04 MDL Number: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Synonym: methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
| PubChem CID | 887 |
|---|---|
| CAS | 67-56-1 |
| Molecular Weight (g/mol) | 32.04 |
| ChEBI | CHEBI:17790 |
| MDL Number | MFCD00004595 |
| SMILES | CO |
| Synonym | methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit |
| IUPAC Name | methanol |
| InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
| Molecular Formula | CH4O |
Methanol (Certified ACS), Fisher Chemical™
CAS: 67-56-1 Molecular Formula: CH4O Molecular Weight (g/mol): 32.04 MDL Number: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Synonym: methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
| PubChem CID | 887 |
|---|---|
| CAS | 67-56-1 |
| Molecular Weight (g/mol) | 32.04 |
| ChEBI | CHEBI:17790 |
| MDL Number | MFCD00004595 |
| SMILES | CO |
| Synonym | methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit |
| IUPAC Name | methanol |
| InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
| Molecular Formula | CH4O |
Methanol (NF), Fisher Chemical™
CAS: 67-56-1 Molecular Formula: CH4O Molecular Weight (g/mol): 32.04 MDL Number: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Synonym: methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
| PubChem CID | 887 |
|---|---|
| CAS | 67-56-1 |
| Molecular Weight (g/mol) | 32.04 |
| ChEBI | CHEBI:17790 |
| MDL Number | MFCD00004595 |
| SMILES | CO |
| Synonym | methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit |
| IUPAC Name | methanol |
| InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
| Molecular Formula | CH4O |
Methanol, ACS Reagent Grade, Absolute, Ricca Chemical
CAS: 67-56-1 Molecular Formula: CH4O Molecular Weight (g/mol): 32.04 MDL Number: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
| PubChem CID | 887 |
|---|---|
| CAS | 67-56-1 |
| Molecular Weight (g/mol) | 32.04 |
| ChEBI | CHEBI:17790 |
| MDL Number | MFCD00004595 |
| SMILES | CO |
| IUPAC Name | methanol |
| InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
| Molecular Formula | CH4O |
Methanol, Anhydrous, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 67-56-1 Molecular Formula: CH4O Molecular Weight (g/mol): 32.04 MDL Number: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Synonym: methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
| PubChem CID | 887 |
|---|---|
| CAS | 67-56-1 |
| Molecular Weight (g/mol) | 32.04 |
| ChEBI | CHEBI:17790 |
| MDL Number | MFCD00004595 |
| SMILES | CO |
| Synonym | methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit |
| IUPAC Name | methanol |
| InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
| Molecular Formula | CH4O |
Methyl Alcohol ACS AR, Macron Fine Chemicals™
CAS: 67-56-1 Molecular Formula: CH4O Molecular Weight (g/mol): 32.04 MDL Number: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Synonym: methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
| PubChem CID | 887 |
|---|---|
| CAS | 67-56-1 |
| Molecular Weight (g/mol) | 32.04 |
| ChEBI | CHEBI:17790 |
| MDL Number | MFCD00004595 |
| SMILES | CO |
| Synonym | methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit |
| IUPAC Name | methanol |
| InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
| Molecular Formula | CH4O |
Methanol, Anhydrous, DriSolv™, Septum-Sealed, MilliporeSigma™
CAS: 67-56-1 Molecular Formula: CH4O Molecular Weight (g/mol): 32.04 MDL Number: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Synonym: methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
| PubChem CID | 887 |
|---|---|
| CAS | 67-56-1 |
| Molecular Weight (g/mol) | 32.04 |
| ChEBI | CHEBI:17790 |
| MDL Number | MFCD00004595 |
| SMILES | CO |
| Synonym | methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit |
| IUPAC Name | methanol |
| InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
| Molecular Formula | CH4O |
Methanol, ACS Reagent, Absolute (Methyl Alcohol Anhydrous), Reagents
CAS: 67-56-1 Molecular Formula: CH4O Molecular Weight (g/mol): 32.04 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Synonym: Methyl Alcohol, Wood Alcohol IUPAC Name: methanol SMILES: CO
| CAS | 67-56-1 |
|---|---|
| Molecular Weight (g/mol) | 32.04 |
| SMILES | CO |
| Synonym | Methyl Alcohol, Wood Alcohol |
| IUPAC Name | methanol |
| InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
| Molecular Formula | CH4O |
Methanol, UHPLC-MS
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Unique signal-to-noise specification for the chemical industry where the solvent quality is linked directly to the sensitivity of the mass spectrometer. | CAS: 67-56-1 | CH4O | 32.04 g/mol
| PubChem CID | 887 |
|---|---|
| CAS | 67-56-1 |
| Molecular Weight (g/mol) | 32.04 |
| ChEBI | CHEBI:17790 |
| MDL Number | MFCD00004595 |
| SMILES | CO |
| Synonym | methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit |
| IUPAC Name | methanol |
| InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
| Molecular Formula | CH4O |
Methanol, For HPLC analysis, MilliporeSigma™
CAS: 67-56-1 Molecular Formula: CH4O Molecular Weight (g/mol): 32.04 MDL Number: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Synonym: methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
| PubChem CID | 887 |
|---|---|
| CAS | 67-56-1 |
| Molecular Weight (g/mol) | 32.04 |
| ChEBI | CHEBI:17790 |
| MDL Number | MFCD00004595 |
| SMILES | CO |
| Synonym | methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit |
| IUPAC Name | methanol |
| InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
| Molecular Formula | CH4O |
HYDRANAL™ - Solvent, Reagent for volumetric two-component KF titration (working medium), Solstice
SureTRACE
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
CAS: 67-56-1
| MDL Number | MFCD00147197 |
|---|
Methanol, LC-MS Grade, 99.8+%
CAS: 67-56-1 Molecular Formula: CH4O Molecular Weight (g/mol): 32.04 MDL Number: MFCD00004595 InChI Key: OKKJLVBELUTLKV-UHFFFAOYSA-N Synonym: methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit PubChem CID: 887 ChEBI: CHEBI:17790 IUPAC Name: methanol SMILES: CO
| PubChem CID | 887 |
|---|---|
| CAS | 67-56-1 |
| Molecular Weight (g/mol) | 32.04 |
| ChEBI | CHEBI:17790 |
| MDL Number | MFCD00004595 |
| SMILES | CO |
| Synonym | methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit |
| IUPAC Name | methanol |
| InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
| Molecular Formula | CH4O |