Learn More
Kamiya Biomedical Apo CIII Reagent, 60mL+20mL/PK
SDP
Supplier:  Kamiya Biomedical KAI006
Encompass_Preferred
K-ASSAY Apo CIII (C3) Assay Reagent. R1: 3 x 20mL (liquid), R2: 1 x 20mL (liquid). Assay Range: 3 - 30 mg/dL. For the quantitative determination of human Apolipoprotein CIII (Apo CII) in serum and plasma by immunoturbidimetric assay. FOR RESEARCH USE ONLY IN THE U.S. NOT FOR DIAGNOSTIC PROCEDURES IN THE U.S. Required Calibrator: K-ASSAY Apo AII/CII/CIII Calibrator (Cat. No. KAI-041C). Recommended Controls: K-ASSAY Apo AII/CII/CIII/E Control (Cat. No. K315C-2M). This assay is intended for use on most popular chemistry analyzers that allow open channel reagents. Please contact KAMIYA BIOMEDICAL COMPANY to check parameter availability for your analyzer.
Catalog No. NC1483332