Learn More
Medchemexpress LLC Kainic acid | 487-79-6 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN2309100MG
Encompass_Preferred
Kainic acid is a potent excitotoxic agent and an agonist for a subtype of ionotropic glutamate receptor. It is an analog of L-glutamate, an ionotropic KA receptor agonist, and induces seizures, damaging hippocampal pyramidal neurons. It is used as an effective tool for studying temporal lobe epilepsy (TLE).
  • Potent excitotoxic agent
  • Agonist for ionotropic glutamate receptor
  • Induces seizures
  • Analog of L-glutamate
  • Ionotropic KA receptor agonist
  • Damages hippocampal pyramidal neurons
  • Used in models for temporal lobe epilepsy (TLE)
Catalog No. 50-004-21645