Learn More
Medchemexpress LLC Kainic acid | 487-79-6 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN230950MG
Encompass_Preferred
Kainic acid is a potent excitotoxic agent that acts as an agonist for a subtype of ionotropic glutamate receptor. It is known to induce seizures and is commonly used in research to study conditions like temporal lobe epilepsy. This product is for research use only and not sold to patients.
  • Potent excitotoxic agent
  • Agonist for ionotropic glutamate receptors
  • Induces seizures in various models
  • Used to create epilepsy models for research
  • High purity (99.95% to 99.96%)
  • Available as a white to off-white solid
  • Stable under recommended storage conditions
  • Detailed solubility information provided for in vitro and in vivo applications
  • Shipping at room temperature
  • Multiple storage conditions for powder and in solvent
Catalog No. 50-004-20958