Learn More
Medchemexpress LLC Kallikrein 2 Antibody (YA2729) | 10 UL

Supplier: Medchemexpress LLC HYP8298410UL
Kallikrein 2 Antibody (YA2729) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting Kallikrein 2. This affinity purified antibody is suitable for Western Blot and Immunocytochemistry/Immunofluorescence applications.
- Host: Rabbit
- Clonality: Recombinant, monoclonal
- Reactivity: Human, mouse, rat
- Immunogen: Synthesized peptide from human KLK2 aa210-261/261
- Isotype: IgG
- Appearance: Liquid
- Formulation: Rabbit IgG in 10mM PBS (pH 7.4), 150mM NaCl, 0.05% BSA, 0.02% NaN3, 50% glycerol
- Storage: -20°C for 1 year; avoid freeze/thaw
- Shipping: Blue ice
- Function: Cleaves Met-Lys and Arg-Ser bonds in kininogen
- Predicted MW: 29 kDa
- Observed MW: 33 kDa
- Application ratios: WB: 1:500-1:1000; ICC/IF: 1:50-1:200
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.