Learn More
Medchemexpress LLC Kallikrein-5 Human 50ug
SDP
Supplier:  Medchemexpress LLC HYP70939A50UG
Encompass_Preferred
Recombinant human Kallikrein-5 (KLK5) produced in HEK293 cells with a C-terminal His tag, supplied lyophilized and intended for research use. The protein corresponds to amino acids V23-S293 (accession Q9Y337), is glycosylated (approximately 35-44 kDa), and is provided as a 50 μg vial suitable for enzymatic assays, protease characterization, and antibody validation.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Includes the mature sequence V23-S293 with native glycosylation.
  • Supplied lyophilized; reconstitute according to datasheet recommendations.
  • Stable when stored at -20°C; aliquot and freeze at -20°C or -80°C for long term.
  • Suitable for enzymatic assays and proteolysis studies.
  • For research use only; not for therapeutic or clinical applications.
Catalog No. 50-001-97076