Learn More
Genetex Inc Kallikrein 7 Monoclonal Antibody [MSVA-707M] HistoMAX™ (GTX04475) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04475
Encompass_Preferred
The Kallikrein 7 antibody [MSVA-707M] HistoMAX™ (GTX04475) is a Mouse Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human skin tissue using GTX04475 Kallikrein 7 antibody [MSVA-707M] HistoMAX. A strong kallikrein 7 staining in the stratum granulosum and the keratinizing superficial layer of the skin.
Catalog No. NC3667782